Product Details

SNP ID
rs8078223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:223472 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTAGGGCGGGTTGAGCTCTTATGC[C/T]AGGCACTTTGCTGGGCCCTTAGTGA
Phenotype
MIM: 604881
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPH3AL PubMed Links

Gene Details

Gene
RPH3AL
Gene Name
rabphilin 3A-like (without C2 domains)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190411.1 Intron NP_001177340.1
NM_001190412.1 Intron NP_001177341.1
NM_001190413.1 Intron NP_001177342.1
NM_006987.3 Intron NP_008918.1
XM_011535364.2 Intron XP_011533666.1
XM_011535365.2 Intron XP_011533667.1
XM_011535366.2 Intron XP_011533668.1
XM_011535367.2 Intron XP_011533669.1
XM_011535368.2 Intron XP_011533670.1
XM_011535369.2 Intron XP_011533671.1
XM_017025367.1 Intron XP_016880856.1
XM_017025368.1 Intron XP_016880857.1
XM_017025369.1 Intron XP_016880858.1
XM_017025370.1 Intron XP_016880859.1
XM_017025371.1 Intron XP_016880860.1

View Full Product Details