Product Details

SNP ID
rs2527785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:43928402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATTGCCAGAGGTGGAATTACAG[A/G]ATCTCTGAGAGTATGACTCTTTTTT
Phenotype
MIM: 610337
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107986791 PubMed Links

Gene Details

Gene
LOC107986791
Gene Name
uncharacterized LOC107986791
There are no transcripts associated with this gene.

Gene
UBE2D4
Gene Name
ubiquitin conjugating enzyme E2 D4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015983.3 Intron NP_057067.1
XM_006715734.3 Intron XP_006715797.1
XM_011515420.2 Intron XP_011513722.1
XM_011515422.2 Intron XP_011513724.1
XM_011515423.2 Intron XP_011513725.1
XM_017012292.1 Intron XP_016867781.1
XM_017012293.1 Intron XP_016867782.1
XM_017012294.1 Intron XP_016867783.1
XM_017012295.1 Intron XP_016867784.1
Gene
URGCP
Gene Name
upregulator of cell proliferation
There are no transcripts associated with this gene.

View Full Product Details