Product Details

SNP ID
rs2912560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55756230 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTCAATGAATGAGCTTCTTGAAC[A/T]TGGATCATATTCATTGATGAGAATA
Phenotype
MIM: 606055
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PCTP PubMed Links

Gene Details

Gene
PCTP
Gene Name
phosphatidylcholine transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102402.2 Intron NP_001095872.1
NM_021213.3 Intron NP_067036.2
XM_005257549.1 Intron XP_005257606.1
XM_005257550.3 Intron XP_005257607.1
XM_017024913.1 Intron XP_016880402.1

View Full Product Details