Product Details

SNP ID
rs60023424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:51154159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCTTTGGGCTTTGTCTCTCTCT[C/T]TTTTTTTTTTTTTGGAAGTTGCAGA
Phenotype
MIM: 156490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NME1 PubMed Links

Gene Details

Gene
NME1
Gene Name
NME/NM23 nucleoside diphosphate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000269.2 Intron NP_000260.1
NM_198175.1 Intron NP_937818.1
Gene
NME1-NME2
Gene Name
NME1-NME2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018136.2 Intron NP_001018146.1

View Full Product Details