Product Details

SNP ID
rs4764341
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10240629 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAACTCACTCCCCAATCAAGGTGA[A/G]TGTGTATCTCCCATGAGTGAGTAAA
Phenotype
MIM: 602894
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KLRD1 PubMed Links

Gene Details

Gene
KLRD1
Gene Name
killer cell lectin like receptor D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114396.1 1847 Intron NP_001107868.1
NM_002262.3 1847 Intron NP_002253.1
NM_007334.2 1847 Intron NP_031360.1
XM_006719067.3 1847 Intron XP_006719130.1
XM_011520650.2 1847 Intron XP_011518952.1
XM_011520651.2 1847 Intron XP_011518953.1
XM_017019285.1 1847 Intron XP_016874774.1
XM_017019286.1 1847 Intron XP_016874775.1
XM_017019287.1 1847 Intron XP_016874776.1
XM_017019288.1 1847 Intron XP_016874777.1
XM_017019289.1 1847 Intron XP_016874778.1
XM_017019290.1 1847 UTR 5 XP_016874779.1
XM_017019291.1 1847 Intron XP_016874780.1

View Full Product Details