Product Details

SNP ID
rs6546870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73669476 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTCAGAGAAGTTTGTTATTACCA[A/G]TCATCTGATGCTTACTTCTGTCAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALMS1P1 PubMed Links

Gene Details

Gene
ALMS1P1
Gene Name
ALMS1, centrosome and basal body associated protein pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details