Product Details

SNP ID
rs3732266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69830316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCCGGCCCGATGCGGGGCGGAC[G/T]GGGGAGCGCCTTGGGAGACGCCAGG
Phenotype
MIM: 106491
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANXA4 PubMed Links

Gene Details

Gene
ANXA4
Gene Name
annexin A4
There are no transcripts associated with this gene.

Gene
GMCL1
Gene Name
germ cell-less, spermatogenesis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178439.4 Intron NP_848526.1
XM_011533033.1 Intron XP_011531335.1
XM_011533034.2 Intron XP_011531336.1
XM_017004704.1 Intron XP_016860193.1
XM_017004705.1 Intron XP_016860194.1

View Full Product Details