Product Details
- SNP ID
-
rs10184422
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:28401253 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTGGGGGTTTAAAAAAAAAAAAAA[A/G]GCTTATCAGAGCCCTGCCCCAAACC
- Phenotype
-
MIM: 601575
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ31356
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4666069,rs75806448] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLJ31356
- Gene Name
- uncharacterized protein FLJ31356
There are no transcripts associated with this gene.
- Gene
- FOSL2
- Gene Name
- FOS like 2, AP-1 transcription factor subunit
View Full Product Details