Product Details
- SNP ID
-
rs3198667
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:24821894 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCTGTGAGTCTGACCACGAGGCGG[A/G]CCCCTTCACCTTGGCTGGGCCTGGT
- Phenotype
-
MIM: 600291
MIM: 611504
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADCY3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76078428] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADCY3
- Gene Name
- adenylate cyclase 3
- Gene
- CENPO
- Gene Name
- centromere protein O
View Full Product Details