Product Details
- SNP ID
-
rs2712030
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:6869020 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTCGGGGTCCTGAGATTTGGTTAC[C/T]ATAGCTGGGCACATGGCAGGATGTG
- Phenotype
-
MIM: 611787
MIM: 607810
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CMPK2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs189411761] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CMPK2
- Gene Name
- cytidine/uridine monophosphate kinase 2
There are no transcripts associated with this gene.
- Gene
- RSAD2
- Gene Name
- radical S-adenosyl methionine domain containing 2
View Full Product Details