Product Details

SNP ID
rs2677743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90904279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGGCAGTCTTCTCGCTCTACCT[C/T]ATGCTGGACCGAGTGCAACACGATC
Phenotype
MIM: 190030 MIM: 600988
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FES PubMed Links

Gene Details

Gene
FES
Gene Name
FES proto-oncogene, tyrosine kinase
There are no transcripts associated with this gene.

Gene
MAN2A2
Gene Name
mannosidase alpha class 2A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320977.1 240 Silent Mutation CTC,CTT L24L NP_001307906.1
NM_006122.3 240 Silent Mutation CTC,CTT L24L NP_006113.2
XM_005254910.3 240 Silent Mutation CTC,CTT L24L XP_005254967.1
XM_011521565.2 240 Intron XP_011519867.1
XM_011521566.2 240 Intron XP_011519868.1
XM_017022184.1 240 Silent Mutation CTC,CTT L24L XP_016877673.1
XM_017022185.1 240 Silent Mutation CTC,CTT L24L XP_016877674.1

View Full Product Details