Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001114357.1 | Intron | NP_001107829.1 | |||
XM_005263019.3 | Intron | XP_005263076.1 | |||
XM_011531989.2 | Intron | XP_011530291.1 | |||
XM_017008236.1 | Intron | XP_016863725.1 | |||
XM_017008237.1 | Intron | XP_016863726.1 | |||
XM_017008238.1 | Intron | XP_016863727.1 | |||
XM_017008239.1 | Intron | XP_016863728.1 | |||
XM_017008240.1 | Intron | XP_016863729.1 | |||
XM_017008241.1 | Intron | XP_016863730.1 | |||
XM_017008242.1 | Intron | XP_016863731.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018359.3 | Intron | NP_060829.2 |