Product Details

SNP ID
rs3733229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82631611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTTTCAGGGTTTACCATGTGCA[A/G]GACATGGTGTCAGCCTCTGTGAGGA
Phenotype
MIM: 608370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCD5 PubMed Links

Gene Details

Gene
SCD5
Gene Name
stearoyl-CoA desaturase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037582.2 Intron NP_001032671.2
NM_024906.2 Intron NP_079182.2

View Full Product Details