Product Details

SNP ID
rs11163245
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:81308383 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGATGACAATTTCATATTTAAAAC[A/G]AGTTTGAGAACAGCACCCTACACAG
Phenotype
MIM: 607018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRL2 PubMed Links

Gene Details

Gene
ADGRL2
Gene Name
adhesion G protein-coupled receptor L2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297704.1 Intron NP_001284633.1
NM_001297705.1 Intron NP_001284634.1
NM_001297706.1 Intron NP_001284635.1
NM_012302.3 Intron NP_036434.1
XM_005270666.4 Intron XP_005270723.1
XM_005270668.2 Intron XP_005270725.1
XM_006710485.3 Intron XP_006710548.1
XM_006710488.2 Intron XP_006710551.1
XM_006710489.3 Intron XP_006710552.1
XM_017000782.1 Intron XP_016856271.1
XM_017000783.1 Intron XP_016856272.1
XM_017000784.1 Intron XP_016856273.1
XM_017000785.1 Intron XP_016856274.1
XM_017000786.1 Intron XP_016856275.1
XM_017000787.1 Intron XP_016856276.1
XM_017000788.1 Intron XP_016856277.1
XM_017000789.1 Intron XP_016856278.1
XM_017000790.1 Intron XP_016856279.1
XM_017000791.1 Intron XP_016856280.1
XM_017000792.1 Intron XP_016856281.1
XM_017000793.1 Intron XP_016856282.1
XM_017000794.1 Intron XP_016856283.1
XM_017000795.1 Intron XP_016856284.1
XM_017000796.1 Intron XP_016856285.1
XM_017000797.1 Intron XP_016856286.1
XM_017000798.1 Intron XP_016856287.1

View Full Product Details