Product Details

SNP ID
rs1045318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:170993892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTGTGGGAGAAGGTATGGGTCA[C/T]TTGCCAGCTGGTGCACATCAAATCG
Phenotype
MIM: 608438
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLK1 PubMed Links

Gene Details

Gene
TLK1
Gene Name
tousled like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136554.1 2770 Missense Mutation AAT,AGT N682S NP_001130026.1
NM_001136555.1 2770 Missense Mutation AAT,AGT N634S NP_001130027.1
NM_012290.4 2770 Missense Mutation AAT,AGT N730S NP_036422.3
XM_011512238.2 2770 Missense Mutation AAT,AGT N667S XP_011510540.1
XM_017005421.1 2770 Missense Mutation AAT,AGT N513S XP_016860910.1
XM_017005422.1 2770 Missense Mutation AAT,AGT N513S XP_016860911.1

View Full Product Details