Product Details

SNP ID
hCV27057783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130656155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGATAGGTTAGATATCAGAGAG[C/T]AAACCTAATCTTAAAAATATAGTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POTEI PubMed Links

Gene Details

Gene
POTEI
Gene Name
POTE ankyrin domain family member I
There are no transcripts associated with this gene.

Gene
POTEJ
Gene Name
POTE ankyrin domain family member J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277083.1 Intron NP_001264012.1

View Full Product Details