Product Details

SNP ID
rs113648981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:427932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACATTTCTGTAAAGCCTGTGCTT[A/C]AAACCCTAGAGTGGTCTGGGAGATG
Phenotype
MIM: 609098
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FBXO25 PubMed Links

Gene Details

Gene
FBXO25
Gene Name
F-box protein 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012173.3 Intron NP_036305.2
NM_183420.1 Intron NP_904356.1
NM_183421.1 Intron NP_904357.1
XM_011534748.2 Intron XP_011533050.1
XM_011534749.1 Intron XP_011533051.1
XM_017013313.1 Intron XP_016868802.1
XM_017013314.1 Intron XP_016868803.1

View Full Product Details