Product Details
- SNP ID
-
hCV27061396
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:431544 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATGAAATAAAAGGTGATACAAAT[A/G]TGTAAATAACAGCCCAGTATCCAGC
- Phenotype
-
MIM: 609098
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO25
PubMed Links
Gene Details
- Gene
- FBXO25
- Gene Name
- F-box protein 25
View Full Product Details