Product Details

SNP ID
rs2790174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:58272171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATCTGTTGTTCGTAAATATTTAG[C/T]GCCTCATAAAGATTTTCTTCAGCCT
Phenotype
MIM: 611932 MIM: 609851
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CISD1 PubMed Links

Gene Details

Gene
CISD1
Gene Name
CDGSH iron sulfur domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018464.4 Intron NP_060934.1
XM_017016409.1 Intron XP_016871898.1
Gene
IPMK
Gene Name
inositol polyphosphate multikinase
There are no transcripts associated with this gene.

View Full Product Details