Product Details

SNP ID
rs12924246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:24833308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGGGAGTGGCAGGTGGGTGTGCA[A/G]TATGATGTATCAAGGAGCCTGGGAA
Phenotype
MIM: 610238 MIM: 610739
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC5A11 PubMed Links

Gene Details

Gene
SLC5A11
Gene Name
solute carrier family 5 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258411.2 Intron NP_001245340.1
NM_001258412.2 Intron NP_001245341.1
NM_001258413.2 Intron NP_001245342.1
NM_001258414.1 Intron NP_001245343.1
NM_052944.4 Intron NP_443176.2
XM_005255080.2 Intron XP_005255137.1
XM_005255082.4 Intron XP_005255139.1
XM_006721015.2 Intron XP_006721078.1
XM_011545725.2 Intron XP_011544027.2
XM_011545728.1 Intron XP_011544030.1
XM_017022900.1 Intron XP_016878389.1
XM_017022901.1 Intron XP_016878390.1
XM_017022902.1 Intron XP_016878391.1
XM_017022903.1 Intron XP_016878392.1
XM_017022904.1 Intron XP_016878393.1
XM_017022905.1 Intron XP_016878394.1
XM_017022906.1 Intron XP_016878395.1
XM_017022907.1 Intron XP_016878396.1
XM_017022908.1 Intron XP_016878397.1
XM_017022909.1 Intron XP_016878398.1
Gene
TNRC6A
Gene Name
trinucleotide repeat containing 6A
There are no transcripts associated with this gene.

View Full Product Details