Product Details
- SNP ID
-
rs11581228
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:119508160 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGAGAGAGGAAAACAATAAGGGG[A/G]AGAGAGAGTCGGGGAGAGAGAGTCG
- Phenotype
-
MIM: 109715
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSD3B1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201834332] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSD3B1
- Gene Name
- hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
View Full Product Details