Product Details
- SNP ID
-
rs4075613
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:38412459 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTAGGTATTATCCATGGTTCAGC[A/G]ATTGTCTGGGGTGGACATTATCTGG
- Phenotype
-
MIM: 607320
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM98C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78834767] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM98C
- Gene Name
- family with sequence similarity 98 member C
There are no transcripts associated with this gene.
- Gene
- RASGRP4
- Gene Name
- RAS guanyl releasing protein 4
View Full Product Details