Product Details
- SNP ID
-
rs7848919
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:6532477 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGAGAAAGGCATTCTTCCGATCAA[A/G]ATGCTTTTATTAGAATACTAATAAA
- Phenotype
-
MIM: 238300
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GLDC
PubMed Links
Gene Details
- Gene
- GLDC
- Gene Name
- glycine decarboxylase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000170.2 |
|
Intron |
|
|
NP_000161.2 |
View Full Product Details