Product Details

SNP ID
hCV27097733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47142697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGAAGAAGACAAGTCGCATGTT[C/T]CAGACGTCCAAAACGGGGTCCTTGT
Phenotype
MIM: 300403 MIM: 300080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFB11 PubMed Links

Gene Details

Gene
NDUFB11
Gene Name
NADH:ubiquinone oxidoreductase subunit B11
There are no transcripts associated with this gene.

Gene
RBM10
Gene Name
RNA binding motif protein 10
There are no transcripts associated with this gene.

View Full Product Details