Product Details

SNP ID
rs1001318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29736178 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTCCAACCAGCTCAAGCCTTTTG[C/T]ACACGGGCTGAAGGAGTCTGGGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABP7 PubMed Links

Gene Details

Gene
CABP7
Gene Name
calcium binding protein 7
There are no transcripts associated with this gene.

Gene
ZMAT5
Gene Name
zinc finger matrin-type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003692.1 Intron NP_001003692.1
NM_001318129.1 Intron NP_001305058.1
NM_019103.2 Intron NP_061976.1

View Full Product Details