Product Details
- SNP ID
-
hCV27099444
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:29885693 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGGGTGTGTTAGCAATCATTTGA[G/T]ACCCGGGTTGTGAACCAGGAAGGGT
- Phenotype
-
MIM: 603558
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MTMR3
PubMed Links
Gene Details
- Gene
- MTMR3
- Gene Name
- myotubularin related protein 3
View Full Product Details