Product Details
- SNP ID
-
rs3909036
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:133325646 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGACCCCGCGTCACCCGGCGGGCT[G/C]GGATTCGCAACCCCCAACCCCGAGG
- Phenotype
-
MIM: 604647
- Polymorphism
- G/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CALY
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147254716] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CALY
- Gene Name
- calcyon neuron specific vesicular protein
View Full Product Details