Product Details

SNP ID
rs7475905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133326507 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAACCATTATTGACAAAGAGATTT[T/A]AAAAAAAAAAGGATGGAAGAAAATA
Phenotype
MIM: 604647
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
CALY PubMed Links

Gene Details

Gene
CALY
Gene Name
calcyon neuron specific vesicular protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321617.1 Intron NP_001308546.1
NM_015722.3 Intron NP_056537.1
XM_005252685.4 Intron XP_005252742.2
XM_011539836.2 Intron XP_011538138.1
XM_011539838.2 Intron XP_011538140.1

View Full Product Details