Product Details

SNP ID
rs2980723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133238142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCTCAAATCTGCCTGCGCCGGA[G/A]AGGACCATGGCCGGTAGGTCCGGGT
Phenotype
MIM: 604130 MIM: 607158
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MIR202 PubMed Links

Gene Details

Gene
MIR202
Gene Name
microRNA 202
There are no transcripts associated with this gene.

Gene
MIR202HG
Gene Name
MIR202 host gene
There are no transcripts associated with this gene.

Gene
UTF1
Gene Name
undifferentiated embryonic cell transcription factor 1
There are no transcripts associated with this gene.

Gene
VENTX
Gene Name
VENT homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014468.3 49 Intron NP_055283.1
XM_017016073.1 49 UTR 5 XP_016871562.1

View Full Product Details