Product Details
- SNP ID
-
rs4538892
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:25844256 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTTTTCCTACAAAGAAAAACAA[A/G]TAGTATCCAAAATATTAAGCATTAA
- Phenotype
-
MIM: 609934
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EBF2
PubMed Links
Gene Details
- Gene
- EBF2
- Gene Name
- early B-cell factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022659.3 |
2346 |
UTR 3 |
|
|
NP_073150.2 |
View Full Product Details