Product Details

SNP ID
rs11213800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111248595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCATCTTAATTGTGTCATTGTCT[A/G]TCACATAACTCAGAATATAAATTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf53 PubMed Links

Gene Details

Gene
C11orf53
Gene Name
chromosome 11 open reading frame 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198498.2 Intron NP_940900.1
XM_011542803.2 Intron XP_011541105.1
XM_011542804.2 Intron XP_011541106.1

View Full Product Details