Product Details

SNP ID
rs1005934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33280514 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTTTTATACCTGTCTCCAGCACC[A/G]CCACCTATGCCTTCTGATGAACTAT
Phenotype
MIM: 604424
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIPK3 PubMed Links

Gene Details

Gene
HIPK3
Gene Name
homeodomain interacting protein kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048200.2 Intron NP_001041665.1
NM_001278162.1 Intron NP_001265091.1
NM_001278163.1 Intron NP_001265092.1
NM_005734.4 Intron NP_005725.3
XM_005252729.2 Intron XP_005252786.1
XM_017017076.1 Intron XP_016872565.1
XM_017017077.1 Intron XP_016872566.1

View Full Product Details