Product Details

SNP ID
rs1802709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33087122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGGGTCCGGAATTATAGCTGCT[A/G]GCTTAGCACGGGAGACATCCTGAAA
Phenotype
MIM: 600367 MIM: 612296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSTF3 PubMed Links

Gene Details

Gene
CSTF3
Gene Name
cleavage stimulation factor subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033505.1 1827 Intron NP_001028677.1
NM_001033506.1 1827 Intron NP_001028678.1
NM_001326.2 1827 Missense Mutation CCA,CTA P554L NP_001317.1
Gene
LINC00294
Gene Name
long intergenic non-protein coding RNA 294
There are no transcripts associated with this gene.

Gene
TCP11L1
Gene Name
t-complex 11 like 1
There are no transcripts associated with this gene.

View Full Product Details