Product Details

SNP ID
rs662894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7788483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGGGGGCCCTGTGGTCCCTGAG[G/T]CTGGCCACGGGGTGTGGAAGGGAGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLEC4GP1 PubMed Links

Gene Details

Gene
CLEC4GP1
Gene Name
C-type lectin domain family 4 member G pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details