Product Details

SNP ID
rs2501079
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:224929520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACACCCCCCTTACCCCCAGCACC[A/G]TCGGTGTCACATCGGGGGCCGCCGC
Phenotype
MIM: 603341
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAH14 PubMed Links

Gene Details

Gene
DNAH14
Gene Name
dynein axonemal heavy chain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145154.1 1098 Intron NP_001138626.1
NM_001373.1 1098 Intron NP_001364.1
NM_144989.2 1098 Intron NP_659426.2
XM_006711735.3 1098 UTR 5 XP_006711798.1
XM_011544057.2 1098 UTR 5 XP_011542359.1
XM_011544058.2 1098 UTR 5 XP_011542360.1
XM_011544059.2 1098 UTR 5 XP_011542361.1
XM_011544061.2 1098 UTR 5 XP_011542363.1
XM_011544062.2 1098 UTR 5 XP_011542364.1
XM_011544063.2 1098 UTR 5 XP_011542365.1
XM_011544064.2 1098 UTR 5 XP_011542366.1
XM_011544065.2 1098 UTR 5 XP_011542367.1
XM_011544066.2 1098 UTR 5 XP_011542368.1
XM_011544067.2 1098 UTR 5 XP_011542369.1
XM_011544069.2 1098 UTR 5 XP_011542371.1
XM_011544070.2 1098 UTR 5 XP_011542372.1
XM_011544071.2 1098 UTR 5 XP_011542373.1
XM_011544072.2 1098 UTR 5 XP_011542374.1
XM_011544073.2 1098 Intron XP_011542375.1
XM_011544074.2 1098 Intron XP_011542376.1
XM_011544076.1 1098 Intron XP_011542378.1
XM_011544079.1 1098 Intron XP_011542381.1
XM_011544081.1 1098 Intron XP_011542383.1
XM_017000295.1 1098 UTR 5 XP_016855784.1
XM_017000296.1 1098 UTR 5 XP_016855785.1
XM_017000297.1 1098 UTR 5 XP_016855786.1
XM_017000298.1 1098 UTR 5 XP_016855787.1
XM_017000299.1 1098 Intron XP_016855788.1
XM_017000300.1 1098 UTR 5 XP_016855789.1

View Full Product Details