Product Details

SNP ID
rs2501072
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:224938176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAGACTTCAAAAGCACTGACAAC[A/C]AAAGCAAAAAGAGATTATTGGATTA
Phenotype
MIM: 603341
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAH14 PubMed Links

Gene Details

Gene
DNAH14
Gene Name
dynein axonemal heavy chain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145154.1 Intron NP_001138626.1
NM_001373.1 Intron NP_001364.1
NM_144989.2 Intron NP_659426.2
XM_006711735.3 Intron XP_006711798.1
XM_011544057.2 Intron XP_011542359.1
XM_011544058.2 Intron XP_011542360.1
XM_011544059.2 Intron XP_011542361.1
XM_011544061.2 Intron XP_011542363.1
XM_011544062.2 Intron XP_011542364.1
XM_011544063.2 Intron XP_011542365.1
XM_011544064.2 Intron XP_011542366.1
XM_011544065.2 Intron XP_011542367.1
XM_011544066.2 Intron XP_011542368.1
XM_011544067.2 Intron XP_011542369.1
XM_011544069.2 Intron XP_011542371.1
XM_011544070.2 Intron XP_011542372.1
XM_011544071.2 Intron XP_011542373.1
XM_011544072.2 Intron XP_011542374.1
XM_011544073.2 Intron XP_011542375.1
XM_011544074.2 Intron XP_011542376.1
XM_011544076.1 Intron XP_011542378.1
XM_011544079.1 Intron XP_011542381.1
XM_011544081.1 Intron XP_011542383.1
XM_017000295.1 Intron XP_016855784.1
XM_017000296.1 Intron XP_016855785.1
XM_017000297.1 Intron XP_016855786.1
XM_017000298.1 Intron XP_016855787.1
XM_017000299.1 Intron XP_016855788.1
XM_017000300.1 Intron XP_016855789.1

View Full Product Details