Product Details
- SNP ID
-
rs12402498
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
26
- Location
-
Chr.1:6613720 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGACTACATTTCCCGACAGGCCTC[C/G]CGGCTCTCCCGCCCTCCCTCCCGAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PHF13
PubMed Links
Gene Details
- Gene
- PHF13
- Gene Name
- PHD finger protein 13
View Full Product Details