Product Details
- SNP ID
-
rs132849
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:45164027 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGTTACAAAATGGCCGCCCGGAG[C/T]GTGTTCGGCGCGGTTCCCCCAGCTG
- Phenotype
-
MIM: 604646
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NUP50
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3788637] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NUP50
- Gene Name
- nucleoporin 50
- Gene
- NUP50-AS1
- Gene Name
- NUP50 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details