Product Details

SNP ID
rs6591593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60494909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTCAAATGCTATTGAGAGTTA[C/T]GTGGTGTTACTCCAAGAGTCACAGC
Phenotype
MIM: 606550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MS4A12 PubMed Links

Gene Details

Gene
MS4A12
Gene Name
membrane spanning 4-domains A12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164470.1 Intron NP_001157942.1
NM_017716.2 Intron NP_060186.2
XM_011545117.2 Intron XP_011543419.1

View Full Product Details