Product Details

SNP ID
rs6010987
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63646322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGGAGGGGGACCAGGTGGGGGA[A/G]CCTCACAGGGGACTTTGGTCTGACT
Phenotype
MIM: 608362
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
STMN3 PubMed Links
Additional Information
For this assay, SNP(s) [rs113042387] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
STMN3
Gene Name
stathmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276310.1 Intron NP_001263239.1
NM_015894.3 Intron NP_056978.2

View Full Product Details