Product Details

SNP ID
rs176964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77720141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATCAACAACAAAAAAACCCAA[A/G]AGGCACTTTTGTAGGACACTGACTC
Phenotype
MIM: 610211 MIM: 603055
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLIRP PubMed Links

Gene Details

Gene
SLIRP
Gene Name
SRA stem-loop interacting RNA binding protein
There are no transcripts associated with this gene.

Gene
SNW1
Gene Name
SNW domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318844.1 Intron NP_001305773.1
NM_012245.2 Intron NP_036377.1
XM_005267413.3 Intron XP_005267470.1

View Full Product Details