Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001312660.1 | 206 | Intron | NP_001299589.1 | ||
NM_145870.2 | 206 | Intron | NP_665877.1 | ||
NM_145871.2 | 206 | Intron | NP_665878.2 | ||
XM_011536670.2 | 206 | UTR 5 | XP_011534972.1 | ||
XM_011536671.2 | 206 | Intron | XP_011534973.1 | ||
XM_017021243.1 | 206 | Intron | XP_016876732.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_013382.5 | 206 | Intron | NP_037514.2 | ||
XM_011536675.2 | 206 | Intron | XP_011534977.1 | ||
XM_011536676.2 | 206 | Intron | XP_011534978.1 | ||
XM_011536677.2 | 206 | Intron | XP_011534979.1 | ||
XM_011536679.2 | 206 | Intron | XP_011534981.1 | ||
XM_017021244.1 | 206 | Intron | XP_016876733.1 | ||
XM_017021245.1 | 206 | Intron | XP_016876734.1 |