Product Details

SNP ID
rs11159268
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77507838 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGGCTTAGTAGCCAAAAAATAAT[A/C]TTTGCTTTTTATACCTCCCCATTCA
Phenotype
MIM: 612684 MIM: 605713
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ISM2 PubMed Links
Additional Information
For this assay, SNP(s) [rs75823402] are located under a probe and SNP(s) [rs78947178] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ISM2
Gene Name
isthmin 2
There are no transcripts associated with this gene.

Gene
SPTLC2
Gene Name
serine palmitoyltransferase long chain base subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004863.3 6323 UTR 3 NP_004854.1
XM_011537384.2 6323 Intron XP_011535686.1

View Full Product Details