Product Details

SNP ID
rs1812642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20558783 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGACACCCAGGTGATTGTGAGGG[A/T]ATTTCCAGTGACCCTGGGCTGATGG
Phenotype
MIM: 603984
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF737 PubMed Links

Gene Details

Gene
ZNF737
Gene Name
zinc finger protein 737
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159293.1 Intron NP_001152765.1
XM_005259697.3 Intron XP_005259754.1
XM_006722597.3 Intron XP_006722660.1
XM_011527598.2 Intron XP_011525900.1
XM_011527599.2 Intron XP_011525901.1
XM_017026116.1 Intron XP_016881605.1
XM_017026117.1 Intron XP_016881606.1

View Full Product Details