Product Details
- SNP ID
-
rs7900
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1938509 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGATTGTGAAACAGGGTTTAGG[A/T]TTCTCCCCTCGTGGACGAGAGGCCA
- Phenotype
-
MIM: 606216
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MSRB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114297845] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MSRB1
- Gene Name
- methionine sulfoxide reductase B1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016332.2 |
1091 |
UTR 3 |
|
|
NP_057416.1 |
- Gene
- RPL3L
- Gene Name
- ribosomal protein L3 like
There are no transcripts associated with this gene.
View Full Product Details