Product Details

SNP ID
rs4360596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14961078 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATAACATGCTCCTCAATTTAAC[C/T]ATGGGATTATGTCCCAATACACCCA
Phenotype
MIM: 605988 MIM: 614174
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCLRE1C PubMed Links

Gene Details

Gene
DCLRE1C
Gene Name
DNA cross-link repair 1C
There are no transcripts associated with this gene.

Gene
MEIG1
Gene Name
meiosis/spermiogenesis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080836.2 Intron NP_001074305.1

View Full Product Details