Product Details
- SNP ID
-
rs12573063
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:13439530 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAATGAATGAGTGAATGAATGAA[C/T]GAATGGAGTAACTCACCTGTCTCCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BEND7
PubMed Links
Gene Details
- Gene
- BEND7
- Gene Name
- BEN domain containing 7
View Full Product Details