Product Details

SNP ID
hCV27229656
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67831476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCATTAATAAATAGTCTTGCAC[A/G]CAGTTCGTGCTTTTCTATTCTGCTC
Phenotype
MIM: 601819
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BPTF PubMed Links

Gene Details

Gene
BPTF
Gene Name
bromodomain PHD finger transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004459.6 Intron NP_004450.3
NM_182641.3 Intron NP_872579.2

View Full Product Details