Product Details

SNP ID
rs2814472
Assay Type
Functionally tested
NCBI dbSNP Submissions
64
Location
Chr.1:165828729 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGCCTGTTTCCTTTGTTTTATC[A/C]GCAAGGCAAGGAGAGACAAGGGAAA
Phenotype
MIM: 609329
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UCK2 PubMed Links

Gene Details

Gene
UCK2
Gene Name
uridine-cytidine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012474.4 Intron NP_036606.2
XM_011509970.2 Intron XP_011508272.2
XM_017002236.1 Intron XP_016857725.1

View Full Product Details