Product Details

SNP ID
rs2119901
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:166606562 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGATTTGTAAGGCCATCAGTAGC[C/T]TTTACGATTGCCTCAGTTCCTGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FMO9P PubMed Links

Gene Details

Gene
FMO9P
Gene Name
flavin containing monooxygenase 9 pseudogene
There are no transcripts associated with this gene.

View Full Product Details